PCT, a 116 amino acid (aa) protein, is comprised of three sections including a 57 aa N-terminal PCT, a 32 aa calcitonin a 21 aa katacalcin. Calcitonin hormone, derived from PCT cleavage. PCT good diagnosis marker for bacterial infection. Other diseases such as sepsis, inflammation, sur, Size: 1mg
Catalog #: 76511-696
Supplier: Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.