ContactUsLinkComponent
PCT, a 116 amino acid (aa) protein, comprised of three sections including a 57 aa N-terminal PCT, a 32 aa calcitonin a 21 aa katacalcin. Calcitonin hormone, derived from PCT cleavage. PCT good diagnos marker for bacterial infection. Other diseases such as sepsis, inflammation, sur, Size: 100mg
Catalog #: 76511-664
Supplier:  Genscript
PCT, a 116 amino acid (aa) protein, comprised of three sections including a 57 aa N-terminal PCT, a 32 aa calcitonin a 21 aa katacalcin. Calcitonin hormone, derived from PCT cleavage. PCT good diagnos marker for bacterial infection. Other diseases such as sepsis, inflammation, sur, Size: 100mg
Catalog #: 76511-664
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.