NGAL (Neutrophil gelatinase-associated lipocalin, also known as lipocalin-2) small protein found in renal tubules. Its expression dramatically increased in ischemic or nephrotoxic injury. It has been implicated in variety of processes including cell differentiation, proliferation, surv, Size: 100mg
Catalog #: 76511-628
Supplier: Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.