ContactUsLinkComponent
D-dimer known as fibrin degradation product, presented in blood after blood clot degraded by fibrinolysis. Its concentration in blood increases when deep venous thrombos (DVT), pulmonary embolism (PE) or disseminated intravascular coagulation (DIC) happens. It serves as useful maker fo, Size: 100mg
Catalog #: 76511-580
Supplier:  Genscript
D-dimer known as fibrin degradation product, presented in blood after blood clot degraded by fibrinolysis. Its concentration in blood increases when deep venous thrombos (DVT), pulmonary embolism (PE) or disseminated intravascular coagulation (DIC) happens. It serves as useful maker fo, Size: 100mg
Catalog #: 76511-580
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.