ContactUsLinkComponent
Creatine kinase (CK) is a dimeric enzyme, including three isoforms of creatine kinase: BB, MM, and MB. Level of serum CK-MB will be increased within 4-6 hours after a myocardial infarction (MI). CK-MB is considered as a useful marker for the diagnosis of myocardial infarction, Size: 1mg
Catalog #: 76511-546
Supplier:  Genscript
Creatine kinase (CK) is a dimeric enzyme, including three isoforms of creatine kinase: BB, MM, and MB. Level of serum CK-MB will be increased within 4-6 hours after a myocardial infarction (MI). CK-MB is considered as a useful marker for the diagnosis of myocardial infarction, Size: 1mg
Catalog #: 76511-546
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.