ContactUsLinkComponent
Lipoprotein-associated phospholipase A2 (LpPLA2) is an enzyme which participates in proinflammatory process. High concentration of LpPLA2 in serum indicates risk of heart attack and stroke. LpPLA2 is considered as specific coronary marker for diagnosing cardiovascular heart disease, Size: 10mg
Catalog #: 76511-514
Supplier:  Genscript
Lipoprotein-associated phospholipase A2 (LpPLA2) is an enzyme which participates in proinflammatory process. High concentration of LpPLA2 in serum indicates risk of heart attack and stroke. LpPLA2 is considered as specific coronary marker for diagnosing cardiovascular heart disease, Size: 10mg
Catalog #: 76511-514
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.