ContactUsLinkComponent
S100 protein family contains variety of members such as BB homodimer AB heterodimer. It useful marker for monitoring brain damage detecting secondary neurological injuries. Concentration of S100 A8/A9 hetero complex increases in blood when special diseases take places, such as rheum, Size: 10mg
Catalog #: 76511-496
Supplier:  Genscript
S100 protein family contains variety of members such as BB homodimer AB heterodimer. It useful marker for monitoring brain damage detecting secondary neurological injuries. Concentration of S100 A8/A9 hetero complex increases in blood when special diseases take places, such as rheum, Size: 10mg
Catalog #: 76511-496
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.