ContactUsLinkComponent
Heart-type fatty acid binding protein (H-FABP) is encoded by FABP3 gene with a low molecular weight of 15 kDa. It is released in large quantities into blood at an early stage of acute myocardial infarction (AMI). H-FABP serum marker with high sensitivity specificity for diagnosing AMI, Size: 1mg
Catalog #: 76511-476
Supplier:  Genscript
Heart-type fatty acid binding protein (H-FABP) is encoded by FABP3 gene with a low molecular weight of 15 kDa. It is released in large quantities into blood at an early stage of acute myocardial infarction (AMI). H-FABP serum marker with high sensitivity specificity for diagnosing AMI, Size: 1mg
Catalog #: 76511-476
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.