ContactUsLinkComponent
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing. The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction. It can be used for diagnosis of congestive heart failure (CHF), Size: 10mg
Catalog #: 76511-430
Supplier:  Genscript
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing. The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction. It can be used for diagnosis of congestive heart failure (CHF), Size: 10mg
Catalog #: 76511-430
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.