Myoglobin (symbol Mb or MB) is an iron-and oxygen-binding protein. Its level in blood increases above normal levels at the beginning of myocardial necrosis associated with myocardial infarction (MI). It is used as a diagnosis marker for skeletal or myocardial muscle injury, Size: 1mg
Catalog #: 76511-380
Supplier: Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.