ContactUsLinkComponent
Cardiac TnT (cTnT) is largest subunit of troponin complex which is composed of cTnT, troponin I (TnI), troponin C (TnC). It is encoded by TNNT2 gene with a calculated molecular weight of 34.6 kDa. CTnT is considered as a useful biomarker for diagnosis of acute myocardial infarcti, Size: 100mg
Catalog #: 76511-372
Supplier:  Genscript
Cardiac TnT (cTnT) is largest subunit of troponin complex which is composed of cTnT, troponin I (TnI), troponin C (TnC). It is encoded by TNNT2 gene with a calculated molecular weight of 34.6 kDa. CTnT is considered as a useful biomarker for diagnosis of acute myocardial infarcti, Size: 100mg
Catalog #: 76511-372
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.