ContactUsLinkComponent
dNTP Mix, 10 mM is suitable for use in PCR, sequencing, nick translation, fill-ins, cDNA synthesis, TdT tailing reactions, and dilution of radiolabeled dNTPs. These nucleotides are supplied as convenient, ready-to-use solutions in sterilized water. DNTP Mix, 10 mM contains all four dNTPs, each
Catalog #: 76510-242
Supplier:  Genscript
dNTP Mix, 10 mM is suitable for use in PCR, sequencing, nick translation, fill-ins, cDNA synthesis, TdT tailing reactions, and dilution of radiolabeled dNTPs. These nucleotides are supplied as convenient, ready-to-use solutions in sterilized water. DNTP Mix, 10 mM contains all four dNTPs, each
Catalog #: 76510-242
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.