ContactUsLinkComponent
Phosphotyrosine Monoclonal Antibody, Host: Mouse, Clone: PY265, CF647 conjugate, Species reactivity: All species, Isotype: IgG1, kappa, Immunogen: Phosphotyrosine conjugated to BSA Synonyms: P-Tyr, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76507-900
Supplier:  Biotium
Phosphotyrosine Monoclonal Antibody, Host: Mouse, Clone: PY265, CF647 conjugate, Species reactivity: All species, Isotype: IgG1, kappa, Immunogen: Phosphotyrosine conjugated to BSA Synonyms: P-Tyr, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76507-900
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.