ContactUsLinkComponent
Testosterone Monoclonal Antibody, Host: Mouse, Clone: 4E1G2, CF514 conjugate, Species reactivity: All species, Isotype: IgG1, kappa, Immunogen: Testosterone 3 CMO conjugated to BSA Synonyms: 17ß-Hydroxy-3-oxo-4-androstene, Concentration: 0.1 mg/mL, Size: 500 uL.
Catalog #: 76507-628
Supplier:  Biotium
Testosterone Monoclonal Antibody, Host: Mouse, Clone: 4E1G2, CF514 conjugate, Species reactivity: All species, Isotype: IgG1, kappa, Immunogen: Testosterone 3 CMO conjugated to BSA Synonyms: 17ß-Hydroxy-3-oxo-4-androstene, Concentration: 0.1 mg/mL, Size: 500 uL.
Catalog #: 76507-628
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.