ContactUsLinkComponent
Pseudomonas Aeruginosa Serotype 6C Monoclonal Antibody, Host: Mouse, Clone: 1200/472, Purified, BSA-free, 1 mg/mL, 50 uL., Species reactivity: Pseudomonas aeruginosa serotype 6C, Isotype: IgG1, kappa, Immunogen: Pseudomonas aeruginosa serotype 6C, Concentration: 1 mg/mL, Size: 50 uL.
Catalog #: 76507-534
Supplier:  Biotium
Pseudomonas Aeruginosa Serotype 6C Monoclonal Antibody, Host: Mouse, Clone: 1200/472, Purified, BSA-free, 1 mg/mL, 50 uL., Species reactivity: Pseudomonas aeruginosa serotype 6C, Isotype: IgG1, kappa, Immunogen: Pseudomonas aeruginosa serotype 6C, Concentration: 1 mg/mL, Size: 50 uL.
Catalog #: 76507-534
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.