ContactUsLinkComponent
Pseudomonas Aeruginosa Serotype 6C Monoclonal Antibody, Host: Mouse, Clone: 1200/472, CF647 conjugate, Species reactivity: Pseudomonas aeruginosa serotype 6C, Isotype: IgG1, kappa, Immunogen: Pseudomonas aeruginosa serotype 6C, Concentration: 0.1 mg/mL, Size: 500 uL.
Catalog #: 76507-510
Supplier:  Biotium
Pseudomonas Aeruginosa Serotype 6C Monoclonal Antibody, Host: Mouse, Clone: 1200/472, CF647 conjugate, Species reactivity: Pseudomonas aeruginosa serotype 6C, Isotype: IgG1, kappa, Immunogen: Pseudomonas aeruginosa serotype 6C, Concentration: 0.1 mg/mL, Size: 500 uL.
Catalog #: 76507-510
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.