ContactUsLinkComponent
Cytochrome c Monoclonal Antibody, Host: Mouse, Clone: 7H8.2C12, CF488A conjugate, Species reactivity: Human, Isotype: IgG2b, kappa, Immunogen: Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76506-186
Supplier:  Biotium
Cytochrome c Monoclonal Antibody, Host: Mouse, Clone: 7H8.2C12, CF488A conjugate, Species reactivity: Human, Isotype: IgG2b, kappa, Immunogen: Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76506-186
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.