ContactUsLinkComponent
Ep-CAM/CD326 Monoclonal Antibody, Host: Mouse, Clone: VU-1D9, CF514 conjugate, Species reactivity: Human, Isotype: IgG1, kappa, Immunogen: Small cell lung carcinoma cells Synonyms: Adenocarcinoma-associated Antigen, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76504-392
Supplier:  Biotium
Ep-CAM/CD326 Monoclonal Antibody, Host: Mouse, Clone: VU-1D9, CF514 conjugate, Species reactivity: Human, Isotype: IgG1, kappa, Immunogen: Small cell lung carcinoma cells Synonyms: Adenocarcinoma-associated Antigen, Concentration: 0.1 mg/mL, Size: 100 uL.
Catalog #: 76504-392
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.