Closures for Headspace Vials, Agilent Technologies
Catalog #: CA76508-220
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Cap Size:20 mm
- Description:Headspace crimp caps without septa
- Material:Aluminium
- Cat. no.:CA76508-220
- Cap Color:Silver