Closures for Headspace Vials, Agilent Technologies
Catalog #: CA76508-156
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Cap Size:20 mm
- Septum:PTFE/silicone
- Description:Headspace crimp caps with septa, certified
- Material:Aluminium
- Cat. no.:CA76508-156
- Cap Color:Aluminium