Gas Testing System, Contact Plate Format, for the presence of microorganisms in compressed air and nitrogen/gas samples, supplied from tanks and pipes under pressure, with calibrated flow meter, stainless steel, aspirating head for Petri plates, and digital timer
Catalog #: CA76509-312
Supplier: HARDY DIAGNOSTICS CA
Customers Who Bought This Also Bought