Remote Head Sample System, For Contact Plates, with tri-clamp connection is completely fabricated in AISI 316 stainless steel, Operator must insert the plate with the culture media into the separated aspiration chamber, Stainless Steel Aspirating Head, contact plate format, 1 tube clamp end
Catalog #: CA76509-276
Supplier: HARDY DIAGNOSTICS CA
Customers Who Bought This Also Bought