Remote Head, Funnel System, Sterilizable, Stainless steel aspirating funnel end clamp with s/s aspirating bell and tube end clamp to connect the system to an Air Sampler, simplifies all the cleaning/sterilization operations, risk of contamination associated with operator activities is reduced
Catalog #: CA76509-268
Supplier: HARDY DIAGNOSTICS CA
Customers Who Bought This Also Bought