Sodium Citrate, Trisodium Salt, Dihydrate, Common Name: Citric Acid, Trisodium Dihydrate, CAS Number: 6132-04-3, Molecular Weight: 294.1, Chemical Formula: C6H5Na3O7. 2H2O, Appearance: White crystalline powder, Assay: >/= 99.0 %, pH (5% Solution): 7.0 - 9.0, Size: 10 Kg
Catalog #: 76509-168
Supplier: RPI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.