Glycerine ACS
Catalog #: 76509-166
Supplier: RPI
CAS Number: 56-81-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:10 L
Specifications
Specification Test Results
- Appearance:Clear liquid
- Acrolein and Glucose:Pass Test
- Assay (GC):≥ 99.5 %
- Chlorinated compounds (as Cl):≤ 0.003 %
- Color, APHA:≤ 10
- Fatty Acid Esters (as Butyric Acid):≤ 0.05 %
- Heavy Metals (as Pb):≤ 2 ppm
- Infrared Spectrum:Conforms to standard
- Neutrality:Pass Test
- Residue after Ignition:≤ 0.005 %
- Substances Darkened By Sulfuric Acid:Pass Test
- Sulfate (SO4):≤ 0.001 %
- Water:≤ 0.5 %
- Country of Origin:Report
- Glycerine ACS:
Specification Test Results
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
:Storage temperature: Room Temperature
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt:
:M.W. 92.09 g/mol
EINECS:200-289-5
:18.6
:290
:177
Storage Temperature:Room Temperature
Customers Who Bought This Also Bought