Phosphate Buffered Saline, 1X Solution, Sterile, 4.5ml Pre-Filled Tubes, Sterile, 36 Tubes per Case, Appearance: Clear and Colorless Solution, commonly used in biological research, it is isotonic and non-toxic to most cells, pH: 7.25 - 7.55, Size: 10Ml
Catalog #: 76509-164
Supplier: RPI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.