ContactUsLinkComponent
Sodium Perchlorate, ACS Grade, Common Name: Perchloric acid, Sodium salt, CAS Number: 7601-89-0, Molecular Weight: 122.44, Chemical Formula: NaClO4, Color: White, Form: Powder or Chunks, Size: 500 Grams
Catalog #: 76509-156
Supplier:  RPI
Sodium Perchlorate, ACS Grade, Common Name: Perchloric acid, Sodium salt, CAS Number: 7601-89-0, Molecular Weight: 122.44, Chemical Formula: NaClO4, Color: White, Form: Powder or Chunks, Size: 500 Grams
Catalog #: 76509-156
Supplier:  RPI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.