ContactUsLinkComponent
D-(-)Salicin, Purity: >/=99.0%, Hplc, Appearance: White to off white powder, CAS Number: 138-52-3, Molecular Weight: 286.3, Chemical Formula: C13H18O7, Solubility: Water, Size: 500G
Catalog #: 76509-146
Supplier:  RPI
D-(-)Salicin, Purity: >/=99.0%, Hplc, Appearance: White to off white powder, CAS Number: 138-52-3, Molecular Weight: 286.3, Chemical Formula: C13H18O7, Solubility: Water, Size: 500G
Catalog #: 76509-146
Supplier:  RPI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.