VWR® Inserts for Wide Opening Vials
Catalog #: 82028-454A
Supplier: VWR International
Specifications
- Cap included:No
- Capacity:0.50 ml
- Color:Clear
- Description:Insert for wide opening vials
- Material:Glass
- Cat. no.:82028-454A
- Bottom Style:Flat bottom