VWR® Screw Caps for 4 ml Screw-Thread Vials
Catalog #: 82028-444A
Supplier: VWR International
Specifications
- Cap Size:13-425
- Septum:Red PTFE/white silicone
- Description:Screw caps with septa
- Material:Phenolic
- Cat. no.:82028-444A
- Cap Color:Black