VWR® Screw Caps for 4 ml Screw-Thread Vials
Catalog #: 82028-442A
Supplier: VWR International
Specifications
- Cap Size:13-425
- Septum:Blue PTFE/white silicone, pre-slit
- Description:Screw caps with septa
- Material:PP
- Cat. no.:82028-442A
- Cap Color:White