VWR® Screw Caps for 10 mm Screw-Thread Vials, Wide Opening
Catalog #: 82028-434A
Supplier: VWR International
Specifications
- Cap Size:10-425
- Septum:Red PTFE/white silicone
- Description:Screw caps with septa
- Material:PP
- Cat. no.:82028-434A
- Cap Color:Black