VWR® Inserts for Wide Opening Vials
Catalog #: 46610-762A
Supplier: VWR International
Specifications
- Cap included:No
- Capacity:0.38 ml
- Color:Clear
- Description:Insert with poly-support spring for wide opening vials
- Material:Glass
- Cat. no.:46610-762A
- Bottom Style:Conical bottom