VWR® Crimp Caps for 11 mm Crimp Top Vials
Catalog #: 46610-754A
Supplier: VWR International
Specifications
- Cap Size:11 mm
- Septum:Clear PTFE/Synthetic Red rubber
- Description:Crimp caps with septa
- Material:Aluminium
- Cat. no.:46610-754A
- Cap Color:Silver
Specifications
About this item
Customers Who Bought This Also Bought
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$45.90 to $336.74
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$386.44 to $1,545.78
Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
Supplier: VWR International
$260.01 to $668.36
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$135.75 to $468.77
Nalgene® 890 Teflon® FEP Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
$276.42 to $310.84
Microliter™ and Gastight® Agilent 7673, 7683, 7693 and 6850 ALS GC Syringes, Hamilton
Supplier: Hamilton
$125.61
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$231.64
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$435.47 to $1,186.77