ContactUsLinkComponent
Oven, 7.5Cf, Across International vacuum oven for optimal mass transfer & acid conversion, can be customized for terpene collection, 5 aluminum heating shelves w/ individual in-shelf temp, sensor, Voltage: 220V, Size: 50/60Hz
Catalog #: CA76501-240
Supplier:  United Science
Oven, 7.5Cf, Across International vacuum oven for optimal mass transfer & acid conversion, can be customized for terpene collection, 5 aluminum heating shelves w/ individual in-shelf temp, sensor, Voltage: 220V, Size: 50/60Hz
Catalog #: CA76501-240
Supplier:  United Science
Customers Who Bought This Also Bought