Chiller, 6100 Series, PolyScience, 10 psi; 15.5 l/min, -10 - 40 degrees, Centrifugal Pump, R134a Refrigerant, Size: 1Hp
Catalog #: CA76501-238
Supplier: United Science
Chiller, 6100 Series, PolyScience, 10 psi; 15.5 l/min, -10 - 40 degrees, Centrifugal Pump, R134a Refrigerant, Size: 1Hp
Catalog #: CA76501-238
Supplier: United Science
Customers Who Bought This Also Bought
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
$73.15
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
$89.92 to $1,797.48
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$771.89
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Supplier: VWR International
$49.22 to $492.14