ContactUsLinkComponent
Evaporator, Rotary, that can process 2 GPH, 60 Hz, Single Phase. 4300 W, High borosilicate glass 3.3, Capacity: 13.2 gallon (50L) evaporating flask, Capacity: 2.64 gallon (10L) dual receiving flask, Welch oil-free vacuum pump, Size: 20L, Voltage: 220V
Catalog #: CA76501-236
Supplier:  United Science
Evaporator, Rotary, that can process 2 GPH, 60 Hz, Single Phase. 4300 W, High borosilicate glass 3.3, Capacity: 13.2 gallon (50L) evaporating flask, Capacity: 2.64 gallon (10L) dual receiving flask, Welch oil-free vacuum pump, Size: 20L, Voltage: 220V
Catalog #: CA76501-236
Supplier:  United Science
Customers Who Bought This Also Bought