ContactUsLinkComponent
Hammer Mill, Pharma Grade, SS, Front removable cover designed to allow full access to the internal grinding chamber w/o disturbing inlet hoppers or transitions, Machined & welded shaft assembly dynamically balanced prior to assembly, Interior product contact surfaces ground to 60-80 grit finish
Catalog #: CA76501-232
Supplier:  United Science
Hammer Mill, Pharma Grade, SS, Front removable cover designed to allow full access to the internal grinding chamber w/o disturbing inlet hoppers or transitions, Machined & welded shaft assembly dynamically balanced prior to assembly, Interior product contact surfaces ground to 60-80 grit finish
Catalog #: CA76501-232
Supplier:  United Science
Customers Who Bought This Also Bought