ContactUsLinkComponent
Cone Mill Grinder, M20 With Preshredder shear Attachment, Stainless Steel, Pharma Grade, engineered for grinding hemp and cannabis, 203 mm Nominal Diameter Screen, 480 RPM Standard Impellow Speed, 5.5 HP Drive Motor, 425 pounds/hr throughput, spare and care kit included
Catalog #: CA76501-230
Supplier:  United Science
Cone Mill Grinder, M20 With Preshredder shear Attachment, Stainless Steel, Pharma Grade, engineered for grinding hemp and cannabis, 203 mm Nominal Diameter Screen, 480 RPM Standard Impellow Speed, 5.5 HP Drive Motor, 425 pounds/hr throughput, spare and care kit included
Catalog #: CA76501-230
Supplier:  United Science
Customers Who Bought This Also Bought