ContactUsLinkComponent
Hammer Mill, 440 lbs./hr. W/ 1/32" screen, 1100 lbs./hr. With 1/16in screen, 4400 RPM w/ Variable Frequency Drive (VFD), Comes with dust bag & holder, material collecting cyclone & duct work connections, voltage: 230V/60hz/3ph
Catalog #: CA76501-228
Supplier:  United Science
Hammer Mill, 440 lbs./hr. W/ 1/32" screen, 1100 lbs./hr. With 1/16in screen, 4400 RPM w/ Variable Frequency Drive (VFD), Comes with dust bag & holder, material collecting cyclone & duct work connections, voltage: 230V/60hz/3ph
Catalog #: CA76501-228
Supplier:  United Science
Customers Who Bought This Also Bought