ContactUsLinkComponent
Heater, For Fractron 100, Delta T Systems Heater, 1 zones of 18 kW of heating capacity, Pump Capacity: 1 HP, 18 GPM at 30 PS, 1/8 DIN Controller - 4-20MA/0--10V, Zone 1, CE Certification available, Requires two 55-gal drums of Paratherm NF oil, voltage: 230 V
Catalog #: CA76501-222
Supplier:  United Science
Heater, For Fractron 100, Delta T Systems Heater, 1 zones of 18 kW of heating capacity, Pump Capacity: 1 HP, 18 GPM at 30 PS, 1/8 DIN Controller - 4-20MA/0--10V, Zone 1, CE Certification available, Requires two 55-gal drums of Paratherm NF oil, voltage: 230 V
Catalog #: CA76501-222
Supplier:  United Science
Customers Who Bought This Also Bought