ContactUsLinkComponent
Cold Trap, Terpene Condenser System, Wall Mount, for easy collection of solvents or terpenes under vacuum, Can be linked to multiple vacuum distillation units, Includes Welch vacuum pump & chiller, 1ph/2YR Warranty/UL/CSA, Voltage: 240V
Catalog #: CA76501-198
Supplier:  United Science
Cold Trap, Terpene Condenser System, Wall Mount, for easy collection of solvents or terpenes under vacuum, Can be linked to multiple vacuum distillation units, Includes Welch vacuum pump & chiller, 1ph/2YR Warranty/UL/CSA, Voltage: 240V
Catalog #: CA76501-198
Supplier:  United Science
Customers Who Bought This Also Bought