ContactUsLinkComponent
Microplate, 1536 Well, TC Treated, Cyclic Olefin Copolymer, tissue culture treated, solid white plate, flat bottom, with PS lid, w/barcode labels, Non-pyrogenic, Length of Plate - 5.030 in, Width of Plate - 3.365 in, Depth of Well - 0.195 in, Working volume: 8ul
Catalog #: 76501-086
Supplier:  Corning
Microplate, 1536 Well, TC Treated, Cyclic Olefin Copolymer, tissue culture treated, solid white plate, flat bottom, with PS lid, w/barcode labels, Non-pyrogenic, Length of Plate - 5.030 in, Width of Plate - 3.365 in, Depth of Well - 0.195 in, Working volume: 8ul
Catalog #: 76501-086
Supplier:  Corning
Customers Who Bought This Also Bought