Circulator, Presto A30 Refrigerated/Heating, 208V, Pump capacity: 25 L/min; 7.25 PSI (Centrifugal pump), Temperature range: -30 to 250C, Heating power: 2.3 kW, Cooling: 500 W 200C, 20 C; 200 W -20 C; 50 W -30 C, Pump port: M24 x 1.5 male
Catalog #: CA10790-514
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought