Chiller, F Series Recirculating, F1000, Fill volume: 7 - 9.5 L, Flow rate: 23 L/m, Temperature range: 0 to 40 deg C, Pump pressure: 14.5 psi, Capacity: 1000W ( 20 deg C); 700W ( 10 deg C); 650W ( 5 deg C); 350W ( 0 deg C)
Catalog #: CA10790-512
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought