Circulator, Presto A80 Refrigerated/Heating, 208V, Pump capacity: 16 to 38 L/min; 1.45 to 18.9 PSI (Adjustable centrifugal pump), Temperature range: -80 to 250C, Heating power: 1.5 kW, Cooling: 1.2 kW 200C, 20C; 1.1 kW -20C,-40C; 650 W -60C; 100 W -80C, Pump port: M24 x 1.5 male
Catalog #: CA10790-458
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought