Circulator, Presto W40 Refrigerated/Heating, 230V, Pump capacity: 16 to 40 L/min; 4.3 to 24.7 PSI (Adjustable centrifugal pump), Temperature range: -40 to 250C, Heating power: 2.8 kW heating, Cooling: 1.2 kW 200C, 20C; 550 W -20C; 60 W -40C, Pump port: M24 x 1.5 male
Catalog #: CA10790-456
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought