Circulator, Presto W40 Refrigerated/Heating, 208V, Pump capacity: 16 to 38 L/min; 1.45 to 18.9 PSI (Adjustable centrifugal pump), Temperature range: -40 to 250C, Heating power: 2.3 kW, Cooling: 1.2 kW 200C, 20C; 550 W -20C; 60 W -40C, Pump port: M24 x 1.5 male
Catalog #: CA10790-454
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought