Heating Circulator, Forte HT60-M3-CU Highly Dynamic, Electrical: 230V, 60Hz, 14A, Temperature range: 70 to 400 deg C, Heating power: 3 kW Heating Power, Pump port: M16 x 1 male
Catalog #: CA10790-446
Supplier: JML SCIENCES INC CA
Heating Circulator, Forte HT60-M3-CU Highly Dynamic, Electrical: 230V, 60Hz, 14A, Temperature range: 70 to 400 deg C, Heating power: 3 kW Heating Power, Pump port: M16 x 1 male
Catalog #: CA10790-446
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
31,17 $ to 282,75 $
VWR® Single Channel Electronic Timer with Memory and Certificate of Calibration
Supplier: VWR International
77,73 $ to 1 082,63 $
VWR® Single Channel Electronic Timer with Memory and Clock and Certificate of Calibration
Supplier: VWR International
75,03 $ to 739,18 $
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
73,15 $
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
89,92 $ to 1 797,48 $