Heating Circulator, Forte HT30-M1-CU Highly Dynamic, Electrical: 3 x 208V, 60Hz, 14A, Temperature range: 40 to 400 deg C, Heating power: 3 kW Heating Power, Pump port: M16 x 1 male
Catalog #: CA10790-444
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought